Learn More
6-Mercaptopurine monohydrate, 98%, Thermo Scientific Chemicals
£53.10 - £217.00
Chemical Identifiers
CAS | 6112-76-1 |
---|---|
Molecular Formula | C5H6N4OS |
Molecular Weight (g/mol) | 170.19 |
MDL Number | MFCD03854445,MFCD01461928 |
InChI Key | WFFQYWAAEWLHJC-UHFFFAOYSA-N |
Synonym | 6-mercaptopurine monohydrate, 6-mercaptopurine hydrate, mercaptopurine monohydrate, mercaptopurine hydrate, 6h-purine-6-thione, 1,7-dihydro-, monohydrate, 6,7-dihydro-3h-purine-6-thione hydrate, purine-6-thiol monohydrate, 1,7-dihydro-6h-purine-6-thione monohydrate, purine-6-thiol, monohydrate, unii-e7wed276i5 |
PubChem CID | 2724350 |
ChEBI | CHEBI:31822 |
IUPAC Name | 3,7-dihydropurine-6-thione;hydrate |
SMILES | O.S=C1N=CNC2=C1NC=N2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10593391
|
Thermo Scientific Acros
226520050 |
5 g | Glass bottle |
£53.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10761501
|
Thermo Scientific Acros
226520250 |
25 g | Glass bottle |
£217.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6112-76-1 | |
170.19 | |
WFFQYWAAEWLHJC-UHFFFAOYSA-N | |
2724350 | |
3,7-dihydropurine-6-thione;hydrate |
C5H6N4OS | |
MFCD03854445,MFCD01461928 | |
6-mercaptopurine monohydrate, 6-mercaptopurine hydrate, mercaptopurine monohydrate, mercaptopurine hydrate, 6h-purine-6-thione, 1,7-dihydro-, monohydrate, 6,7-dihydro-3h-purine-6-thione hydrate, purine-6-thiol monohydrate, 1,7-dihydro-6h-purine-6-thione monohydrate, purine-6-thiol, monohydrate, unii-e7wed276i5 | |
CHEBI:31822 | |
O.S=C1N=CNC2=C1NC=N2 |
Specifications
6112-76-1 | |
100.0 | |
Yellow | |
99.5% min. (HPLC) | |
MFCD03854445,MFCD01461928 | |
6-mercaptopurine monohydrate, 6-mercaptopurine hydrate, mercaptopurine monohydrate, mercaptopurine hydrate, 6h-purine-6-thione, 1,7-dihydro-, monohydrate, 6,7-dihydro-3h-purine-6-thione hydrate, purine-6-thiol monohydrate, 1,7-dihydro-6h-purine-6-thione monohydrate, purine-6-thiol, monohydrate, unii-e7wed276i5 | |
WFFQYWAAEWLHJC-UHFFFAOYSA-N | |
3,7-dihydropurine-6-thione;hydrate | |
2724350 | |
170.19 | |
Fine Powder |
99.5 | |
313.0°C | |
Authentic | |
C5H6N4OS | |
10, 5702 | |
Solubility in water: insoluble. Other solubilities: soluble in diluted alkali solutions,slightly soluble in diluted sulfuric acid,soluble in hot alcohol,insoluble in acetone and ether | |
O.S=C1N=CNC2=C1NC=N2 | |
170.19 | |
CHEBI:31822 | |
99.5+% | |
6-Mercaptopurine monohydrate, 99.5+% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
missing translation for 'rtecsNumber' : UP0400000
- 6-Mercaptopurine monohydrate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only