Learn More
Ohaus™ 6 x Centrifuge Tube Adapters, Open Bottom, Frontier™ Fixed Angle Rotors
Facilitate centrifugation of tubes smaller than the original, provided carrier space for an array of Frontier™ fixed angle rotors.
Brand:  Ohaus™ 30130884
39.49 GBP valid until 2024-11-30
Use promo code "21681" to get your promotional price.
 View more versions of this product
Product Code. 15836632
  • £46.15 / Pack of 6
Explore more special offers
Description
Description
  • Accommodates centrifuge tubes that are smaller than the space provided in the rotor carrier
  • Flat-, open-bottom carrier.
  • Suitable for use in the following Frontier™ rotors:
    • R-A30x2/13MS
    • R-A24x2/15MS
    • R-A24x2/15M
    • R-A12x2/18MI
    • R-A24x2/16MI
    • R-A30x2/14MIS
    • R-A44x2/16MI
    • R-A24x2/16MIS
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.2/0.4 mL
Frontier Fixed-Angle Rotors
1
6 mm
Adapter, D6mm, 0.2/0.4ml Tubes x6
6