Learn More
Ohaus™ 6 x Centrifuge Tube Adapters, Open Bottom, Frontier™ Fixed Angle Rotors
Facilitate centrifugation of tubes smaller than the original, provided carrier space for an array of Frontier™ fixed angle rotors.
£42.50 - £83.10
Specifications
For Use With (Equipment) | Frontier Fixed-Angle Rotors |
---|---|
No. of Places | 1 |
No. per Pack | 6 |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
15836632
|
Ohaus™
30130884 |
0.2/0.4 mL |
£42.50
Pack of 6 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15846632
|
Ohaus™
30130885 |
0.5 mL |
£42.50
Pack of 6 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15856632
|
Ohaus™
30130886 |
1.5/2.0 mL |
£83.10
Pack of 6 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Accommodates centrifuge tubes that are smaller than the space provided in the rotor carrier
- Flat-, open-bottom carrier.
- Suitable for use in the following Frontier™ rotors:
- R-A30x2/13MS
- R-A24x2/15MS
- R-A24x2/15M
- R-A12x2/18MI
- R-A24x2/16MI
- R-A30x2/14MIS
- R-A44x2/16MI
- R-A24x2/16MIS
Specifications
Frontier Fixed-Angle Rotors | |
1 | |
6 |