missing translation for 'onlineSavingsMsg'
Learn More

BD Biosciences™ 647 EdU Click Proliferation Kit (RUO)

For analyzing DNA synthesis in cycling cells.

Brand:  BD Biosciences™ 565456

Additional Details : Weight : 0.19000kg


Product Code. 15874329

  • £890.00 / Each
Estimated Shipment: 03-09-2024
to see stock.
Add to basket

Description

Description

  • Provides a tool for analyzing DNA synthesis in cycling cells.
  • In this method, the thymidine analog EdU (5-ethynyl-2´-deoxyuridine) is incorporated into newly synthesized DNA by cells progressing through the S (DNA synthesis) phase of the cell cycle.
  • EdU can then be detected in fixed and permeabilized cells with a fluorophore-labeled azide.
  • The reaction of the fluorophore-labeled azide and EdU occurs via a copper-catalyzed click reaction between the azide moiety of the fluorophore and the alkyne moiety of EdU.
  • When co-stained with a DNA dye such as 7-AAD, PI, or DAPI, cell populations may be segmented by flow cytometry into the G0/G1-phases (2N DNA content, EdU-negative), S-phase (2N-4N DNA content, EdU-positive), or G2/M-phases (4N DNA content, EdU-negative).
  • Contains Eterneon™ Red 645 Azide, which is excited by the red laser and has an excitation maximum of 643 nm and an emission maximum of 662 nm.
Specifications

Specifications

EdU, Eterneon™ Red 645 Azide, Buffer Additive (10X) to be stored dry and protected from light at -20°C. Saponin-based Permeabilization and Wash Reagent (10X), Fixative, Catalyst, DMSO to be stored dry at 2 - 8°C or room temperature.
50 Tests
Flow Cytometry, Fluorophore
10 mg EdU, 130 μL Eterneon™ Red 645 azide (10 mM), 400 mg Buffer Additive (10X), 50 mL Saponin-based Permeabilization and Wash Reagent (10X), 5 mL Fixative, 2 mL Catalyst, 5 mL DMSO.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 647 EdU Click Proliferation Kit

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : formaldehyde

SDS
Documents

Documents

Certificates
Special Offers

Special Offers