missing translation for 'onlineSavingsMsg'
Learn More

Tuttnauer 65L Benchtop Autoclave

The 65L Benchtop Autoclave has an advanced multi-color control panel and a chamber made of 316L stainless steel. Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, biohazard and waste sterilization, Fo control, and more.

Brand:  Tuttnauer 3850ELC-230-D

 View more versions of this product

Product Code. 17821077

  • £17965.00 / Each
Estimated Shipment: 09-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

EL - D Line models:

Tuttnauer benchtop laboratory autoclaves are front loading autoclaves available in chamber sizes from 28 to 160 liters. EL models have an advanced multi-color control panel and a chamber made of 316L stainless steel. Tuttnauer laboratory autoclaves have been designed to provide high quality repeatable performance and accountability for a wide range of applications used in modern laboratories.

  • Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, biohazard and waste sterilization, Fo control, and more.
  • Benchtop lab autoclaves are designed to save space on your laboratory workbench.
Specifications

Specifications

14.3 gal.
50 cm
20 in.
30.6 in.
3850ELC-D
221°F to 279°F
Cooling
Non-sterile
65 L
15.2 in.
38 cm
765 mm
Benchtop Autoclave
105°C to 137°C
316L Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.