Learn More
Tuttnauer 65L Benchtop Autoclave
The 65L Benchtop Autoclave has an advanced multi-color control panel and a chamber made of 316L stainless steel. Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, biohazard and waste sterilization, Fo control, and more.
£13250.00 - £16371.70
Specifications
Capacity (English) | 14.3 gal. |
---|---|
Capacity (Metric) | 65 L |
Depth (Metric) Chamber | 50 cm |
Diameter (English) Chamber | 15.2 in. |
Depth (English) Chamber | 20 in. |
Product Code | Brand | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | |||||
17831087
|
Tuttnauer
3850EL-230-D |
3850EL-D |
£14598.32
Each |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||||
17821077
|
Tuttnauer
3850ELC-230-D |
3850ELC-D |
£16371.70
Each |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||||
17811087
|
Tuttnauer
3850EAP-D |
3850EAP-D |
£13250.00
Each |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||||
Description
EL - D Line models:
Tuttnauer benchtop laboratory autoclaves are front loading autoclaves available in chamber sizes from 28 to 160 liters. EL models have an advanced multi-color control panel and a chamber made of 316L stainless steel. Tuttnauer laboratory autoclaves have been designed to provide high quality repeatable performance and accountability for a wide range of applications used in modern laboratories.
- Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, biohazard and waste sterilization, Fo control, and more.
- Benchtop lab autoclaves are designed to save space on your laboratory workbench.
Specifications
14.3 gal. | |
50 cm | |
20 in. | |
30.6 in. | |
Benchtop Autoclave | |
105°C to 137°C | |
Non-sterile |
65 L | |
15.2 in. | |
38 cm | |
765 mm | |
221°F to 279°F | |
316L Stainless Steel |