missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ 7960 Series Thermal Transfer Printer Ribbon
Thermal transfer ribbon with excellent smearing resistance and good solvent resistance.
Brand: Brady™
48.64 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
When printed on the appropriate Brady™ label, this ribbon gives good solvent resistance.
- Lower for alcohol
- High print quality
- Excellent smear resistance
Specifications
Black | |
Resin | |
3.3 in. | |
85 mm | |
12.7 mm | |
2011/65/EU and 2015/863 |
BBP11 Label Printer, BBP12 Label Printer | |
230 ft. | |
70 m | |
0.5 in. | |
Open-core Ribbon |