Learn More
8-Octanolactam, 98%, Thermo Scientific Chemicals
£57.60 - £624.00
Chemical Identifiers
CAS | 935-30-8 |
---|---|
Molecular Formula | C8H15NO |
Molecular Weight (g/mol) | 141.214 |
MDL Number | MFCD00003273 |
InChI Key | YDLSUFFXJYEVHW-UHFFFAOYSA-N |
Synonym | 2-azacyclononanone, caprylolactam, capryllactam, azacyclononan-2-one, 8-octanelactam, 8-aminooctanoic acid lactam, 2-perhydroazoninone, 2h-azonin-2-one, octahydro, cyclooctanone lactam, octamethylenimine, 2-oxo |
PubChem CID | 13632 |
IUPAC Name | azonan-2-one |
SMILES | C1CCCC(=O)NCCC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11428557
|
Thermo Scientific Alfa Aesar
A18621.06 |
5 g |
£57.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11438557
|
Thermo Scientific Alfa Aesar
A18621.14 |
25 g |
£204.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11448557
|
Thermo Scientific Alfa Aesar
A18621.22 |
100 g |
£624.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Azacyclononanone has been used as an internal standard liquid chromatography-ultraviolet detection method for the determination of caprolactam in the aqueous food stimulant 3% acetic acid (w/v).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Azacyclononanone has been used as an internal standard liquid chromatography-ultraviolet detection method for the determination of caprolactam in the aqueous food stimulant 3% acetic acid (w/v).
Solubility
Soluble in methanol (almost transparency).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
935-30-8 | |
141.214 | |
YDLSUFFXJYEVHW-UHFFFAOYSA-N | |
13632 | |
C1CCCC(=O)NCCC1 |
C8H15NO | |
MFCD00003273 | |
2-azacyclononanone, caprylolactam, capryllactam, azacyclononan-2-one, 8-octanelactam, 8-aminooctanoic acid lactam, 2-perhydroazoninone, 2h-azonin-2-one, octahydro, cyclooctanone lactam, octamethylenimine, 2-oxo | |
azonan-2-one |
Specifications
935-30-8 | |
C8H15NO | |
109775 | |
Soluble in methanol (almost transparency). | |
C1CCCC(=O)NCCC1 | |
141.214 | |
141.21 | |
8-Octanolactam |
72°C to 75°C | |
MFCD00003273 | |
2-azacyclononanone, caprylolactam, capryllactam, azacyclononan-2-one, 8-octanelactam, 8-aminooctanoic acid lactam, 2-perhydroazoninone, 2h-azonin-2-one, octahydro, cyclooctanone lactam, octamethylenimine, 2-oxo | |
YDLSUFFXJYEVHW-UHFFFAOYSA-N | |
azonan-2-one | |
13632 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 213-300-3
missing translation for 'rtecsNumber' : CO1757000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only