Learn More
Thermo Scientific™ 8 x 50 Individually Sealed Fixed Angle Rotor for 50 mL conical (for compact and 1.6L General Purpose Centrifuges)
Centrifuge eight conical tubes in individually sealed vessels.
Brand:  Thermo Scientific™ 75003694
1482.54 GBP valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Product Code. 10605835
  • £1830.80 / Each
Explore more special offers
Description
Description
Centrifuge eight conical tubes in individually sealed vessels using Thermo Scientific™ 8 x 50mL Sealed Fixed-Angle Rotor.
Individual sealed canisters for high-risk samples, certified by Public Health England, Porton Down, UK.
Secure, push-button Auto-Lock rotor exchange in as little as 3 seconds delivers:
  • Trouble-free rotor installation and removal
  • No tools are required
  • No complicated rotor changing procedures
  • Clear chamber access for cleaning convenience
  • Quick switch between applications, from conical tubes to blood tubes, from microtubes to microplates, and the flexibility to evolve with the changing needs of your laboratory
TRUSTED_SUSTAINABILITY
Specifications
Specifications
8 x 50mL Individually Sealed Fixed Angle Rotor
8 x 50 mL
6,500 rpm
116 mm
Thermo Scientific Sorvall Legend X1, Sorvall ST 16, Multifuge X1, Megafuge 16 and SL 16 Centrifuges
Fixed Angle Rotor
6,755 x g
143 mm
29.5 mm
25°
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ 8 x 50 Individually Sealed Fixed Angle Rotor for 50 mL conical (for compact and 1.6L General Purpose Centrifuges) > 8 x 50mL Fixed Angle rotor