missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human IL-16 Protein
Recombinant Protein
Brand: Novus Biologicals™ NBP2-35029-100ug
784.80 GBP valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
IL16 Protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using human peripheral T Lymphocytes is in a concentration range of 50-500 ng/mL.
Specifications
Q14005, Q14005, Q14005 | |
PAGE, Bioactivity | |
3603 | |
100 μg | |
Less than 1 EU/ug of IL-16, 121 a.a. as determined by LAL method. | |
IL16 | |
Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity | |
IL-16 |
LYOPH | |
Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. | |
12.4 kDa | |
Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. | |
FLJ16806, FLJ42735, FLJ44234, HsT19289, IL-16, interleukin 16 (lymphocyte chemoattractant factor), LCFprIL-16, lymphocyte chemoattractant factor, neuronal interleukin 16, NIL16, PRIL16, prointerleukin 16, pro-interleukin-16 | |
Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C. | |
>95% by SDS-PAGE and HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Recombinant Human IL-16 Protein > 100 μg
Spot an opportunity for improvement?Share a Content Correction