Learn More
Agilent BioTek 800 TS Absorbance Reader, IVD

The BioTek 800 TS absorbance reader is an affordable, high-quality microplate reader for assays in 6- to 384-well formats. For In Vitro Diagnostic Use.
Brand: Agilent BioTek 800TSNB-SI
6805.80 GBP valid until 2025-12-31
Use promo code "24302" to get your promotional price.
Description
The BioTek 800 TS absorbance reader is an affordable, high-quality microplate reader for assays in 6- to 384-well formats. Applications include ELISA, protein quantification, enzyme kinetics and cell-based assays
The color touch screen provides a visual user interface, making programming fast and intuitive. Expanded functionality and data analysis is available with Gen5 software. The 800 TS partners with the 50 TS washer, making a compact, versatile, and affordable system to automate many application workflows.

Specifications
Microplate Reader | |
USB | |
Endpoint, Kinetic, Well-area Scanning | |
Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option) | |
CE and TUV marked. RoHS compliant. | |
Measuring absorbance in microplate-based assays | |
800TSNB | |
400 to 750 nm | |
8 kg |
Photodiode | |
Tungsten Halogen Lamp | |
Linear (except NB configurations) | |
10 nm | |
405, 450, 490, 630 nm | |
6- to 384-well Terasaki plates; 400 to 750 nm | |
100/240 V | |
18.5 lb. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.