Learn More
Agilent BioTek 800 TS Absorbance Reader, IVD

The BioTek 800 TS absorbance reader is an affordable, high-quality microplate reader for assays in 6- to 384-well formats. For In Vitro Diagnostic Use.
£6235.00 - £8276.00
Specifications
Product Type | Microplate Reader |
---|---|
Detector Type | Photodiode |
Interface | USB |
Light Source | Tungsten Halogen Lamp |
Reading Options | Endpoint, Kinetic, Well-area Scanning |
Product Code | Brand | Model | Filters | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Filters | Includes | Price | Quantity & Availability | |||
17207799
![]() |
Agilent BioTek
800TS-SI |
800TS | 405, 450, 490, 630 nm | 6- to 96-well plates; 400 to 750 nm; Shaking |
£6235.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17227799
![]() |
Agilent BioTek
800TSI-SI |
800TSI | 405, 450, 490, 630 nm | 6- to 96-well plates; 400 to 750 nm; Shaking; Incubation |
£7938.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17247799
![]() |
Agilent BioTek
800TSNB-SI |
800TSNB | 405, 450, 490, 630 nm | 6- to 384-well Terasaki plates; 400 to 750 nm |
£7164.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17267799
![]() |
Agilent BioTek
800TSUV-SI |
800TSUV | 340, 405, 450, 490, 630 nm | 6- to 96-well plates; 340 to 750 nm; Shaking |
£6573.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17287799
![]() |
Agilent BioTek
800TSUVI-SI |
800TSUVI | 340, 405, 450, 490, 630 nm | 6- to 96-well plates; 340 to 750 nm; Shaking; Incubation |
£8276.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
The BioTek 800 TS absorbance reader is an affordable, high-quality microplate reader for assays in 6- to 384-well formats. Applications include ELISA, protein quantification, enzyme kinetics and cell-based assays
The color touch screen provides a visual user interface, making programming fast and intuitive. Expanded functionality and data analysis is available with Gen5 software. The 800 TS partners with the 50 TS washer, making a compact, versatile, and affordable system to automate many application workflows.
Specifications
Microplate Reader | |
USB | |
Endpoint, Kinetic, Well-area Scanning | |
Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option) | |
CE and TUV marked. RoHS compliant. | |
100/240 V | |
8 kg |
Photodiode | |
Tungsten Halogen Lamp | |
Linear (except NB configurations) | |
10 nm | |
Measuring absorbance in microplate-based assays | |
18.5 lb. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.