Learn More
Cole-Parmer™ Adapters and Clamps for High-Energy Mixer/Mill™ Grinders
Use adapters to run multiple samples or to hold vial securely in place during grinding.
Brand:  Cole-Parmer™ 8010
Code : A0
Additional Details : Weight : 2.00000kg
 View more versions of this product
Product Code. 12564236
  • £181.00 / Each
Estimated Shipment: 14-08-2024
to see stock.
Add to basket
Description
Description
  • Suitable for use with the Cole-Parmer™ Freezer/Mill™ Cryogenic Grinders
  • Optimize performance of your Cole-Parmer™ Freezer/Mill™ grinder with a wide range of adapters and clamps.
Specifications
Specifications
Multiple Sample Adapter
Ideal for High Throughput Applications That Involve Processing Multiple Smaller Samples
8000 D and 8000 M Mixer or Mill
7 x 0.5 in. (12.7 mm) dia. Vials
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products