Learn More
Cole-Parmer™ Adapters and Clamps for High-Energy Mixer/Mill™ Grinders
Use adapters to run multiple samples or to hold vial securely in place during grinding.
Brand: Cole-Parmer™ 8012-SSP
Code : NEW
Additional Details : Weight : 2.00000kg
Description
- Suitable for use with the Cole-Parmer™ Freezer/Mill™ Cryogenic Grinders
- Optimize performance of your Cole-Parmer™ Freezer/Mill™ grinder with a wide range of adapters and clamps.
Specifications
Vial Clamp Adapter | |
8000 D and 8000 M Mixer or Mill | |
Plastic |
Ideal for High Throughput Applications That Involve Processing Multiple Smaller Samples | |
Vial Adapters (04576-65 and 04576-66) |