missing translation for 'onlineSavingsMsg'
Learn More

Uvex 95980 Insole For Uvex Naturform - Width 11

Comfortable climatic insole with antistatic properties for Uvex Performance (Naturform)

£15.10

Specifications

Color Black
For Use With (Equipment) Uvex Naturform
Material Microvelour
Disposable Reusable
Gender Unisex
View More Specs

Products 11 Description

Description

  • Full-surface foot shock absorption
  • Highly breathable and moisture-absorbent material
  • Shock absorption zones support natural foot movement
  • Skin compatibility and pleasant surface structure
  • Supports foot arch
  • Article No: 9598.0
  • Sizes: 38 to 48
Specifications

Specifications

Black
Microvelour
Unisex
Yes
Uvex Naturform
Reusable
Standard width
No
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Uvex 95980 Insole For Uvex Naturform - Width 11

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.