Learn More
Eppendorf™ 96-Well Flat Bottom PCR Microplates
For quantitative protein analysis
Brand: Eppendorf™ 0030601475
This item is not returnable.
View return policy
Description
Specifications
Yes | |
Quantitative Protein Analysis | |
80 Plates (5 bags of 16 plates) | |
Polypropylene | |
Flat Bottom (F-well) | |
For PCR techniques |
Standard | |
8 x 12 | |
Gray | |
96 | |
Yes | |
121°C |