Learn More
Fisherbrand™ 96-Well Low-Profile, Skirted PCR Plates, Natural
Fit most thermal cyclers
Brand: Fisherbrand™
35.84 GBP valid until 2024-07-31
Use promo code "21640" to get your promotional price.
Code : LC
Additional Details : Weight : 0.08550kg
Description
Fit most thermal cyclers
- Virgin polypropylene
- Clear well bottom for sample visibility
- Stackable
- Raised-rim design
- Sealable using adhesive films and foils, heat seals, and cap strips
- 200μL maximum well volume when used with adhesive and heat seals
- Cut-away corner and alphanumeric grid
- SBS (Society for Biomolecular Screening) footprint
- Automation friendly
- Eight holes in the skirt aid plate positioning and removal from thermal cycler block
- Cleanroom produced
Specifications
Standard | |
Natural | |
Skirted | |
200 μL | |
PCR, RT-PCR applications |
8 x 12 | |
PCR Microplate | |
Polypropylene | |
96 | |
Yes |