missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 96-well Medium Binding Standard ELISA Microplates
For diagnostics and immunological research applications. Greiner Bio-One 96-Well Medium Binding Standard ELISA Microplates is free of detectable DNase, RNase, human DNA.
£64.99
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Binding Type | Medium Binding |
Color | Clear |
Material | Polystyrene |
Description
- Non-Pyrogenic
- Free of detectable DNase, RNase, human DNA
- Material: Crystal clear polystyrene
- Alphanumeric well coding
- Suitable for transmission measurements
- Well-to-well spacing: 9mm
- Suitable for tra
Certifications
DIN ISO 9001
Specifications
Standard | |
Medium Binding | |
Polystyrene | |
Non-sterile | |
40/Cs. |
8 x 12 | |
Clear | |
96 | |
Microlon 200 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Greiner Bio-One 96-well Medium Binding Standard ELISA Microplates