Learn More
Greiner Bio-One 96-well Medium Binding Standard ELISA Microplates
For diagnostics and immunological research applications. Greiner Bio-One 96-Well Medium Binding Standard ELISA Microplates is free of detectable DNase, RNase, human DNA.
Brand:  Greiner Bio-One 651001
 View more versions of this product
Product Code. 10324822
  • £64.99 / Pack of 40
Estimated Shipment: 10-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Non-Pyrogenic
  • Free of detectable DNase, RNase, human DNA
  • Material: Crystal clear polystyrene
  • Alphanumeric well coding
  • Suitable for transmission measurements
  • Well-to-well spacing: 9mm
  • Suitable for tra
    Certifications
    DIN ISO 9001
    TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
Inner bags of 10
Clear
96
Microlon 200
40/Cs.
8 x 12
Medium Binding
Polystyrene
Non-sterile
Conical Bottom (V-well)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Greiner Bio-One 96-well Medium Binding Standard ELISA Microplates > V-bottom (Conical)