missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 96 Well Black Plate, Non-Treated Surface, No Lid, Non-Sterile, Pack of 10
BIOPHARMA

Ensure consistent and reliable results using black and white plates optimized for robotics, plate readers and liquid handling systems.

Brand:  Thermo Scientific™ 237108

348.64 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 10307451

  • £633.00 / Pack of 180
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Ensure consistent and reliable results using the high quality Thermo Scientific™ Nunc™ F96 MicroWell™ Black and White Polystyrene Plate. Black and white plates are optimized for robotics, plate readers and liquid handling systems and designed for key imaging characteristics. Available with or without cell culture treatment.

  • Background, autoflorescence and crosstalk are improved when you use Nunc Black or White microplates
  • Black microplates are recommended for florescence measurements; with minimum back-scattered light and background fluorescence
  • White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
  • Working volume range: 50 to 250μL/well
  • Choice of Thermo Scientific™ Nunclon™ Delta surface for cell-based assays
  • Customized bar coding is available
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Custom Barcoding Available
Standard
8 x 12
Nunc
Microplate
96
Non-sterile
Flat Bottom (F-well)
10
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Without Lid
10 Pk.
Black
Polystyrene
50 to 250 μL
Non-treated
180
Case of 180
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number184920 Certificate TypeCertificate of Compliance Date05/03/2025 Catalog Number237108
Lot Number184895 Certificate TypeCertificate of Compliance Date23/12/2024 Catalog Number237108
Lot Number184289 Certificate TypeCertificate of Compliance Date13/12/2024 Catalog Number237108
Lot Number179598 Certificate TypeCertificate of Compliance Date06/12/2024 Catalog Number237108
Lot Number165325 Certificate TypeCertificate of Compliance Date06/12/2024 Catalog Number237108
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ 96 Well Black Plate, Non-Treated Surface, No Lid, Non-Sterile, Pack of 10 > 96W Black Polystyrene,Non-Treated, without lid, Non-sterile, 10/180

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.