Learn More
Thermo Scientific™ WebSeal™ 96-Well Plate Kits with Sealing Mats
Save time and minimize the risk of cross-contamination with convenient 96-Well Plate Kits packaged with pre-assembled glass or PTFE vials.
£61.02 - £425.99
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Product Line | Webseal |
Product Type | Glass Inserted Plate Kit with 96-Position Mat |
No. of Wells | 96 |
Product Code | Brand | Total Volume (Metric) Well | Profile | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Total Volume (Metric) Well | Profile | Includes | Price | Quantity & Availability | |||||
15132769
|
Thermo Scientific™
60180-K100 |
500 μL | Taper | Vial, Mat |
£381.90
Pack of 5 |
In Stock
Log in to see stock. |
|||||
15142769
|
Thermo Scientific™
60180-K101 |
700 μL | Taper | Vial, Mat, Cutting Tool |
£325.62
Pack of 5 |
In Stock
Log in to see stock. |
|||||
15152769
|
Thermo Scientific™
60180-K103 |
700 μL | Round | Vial, Mat, Cutting Tool | N/A | ||||||
15162769
|
Thermo Scientific™
60180-K104 |
1100 μL | Round | Vial, Mat, Cutting Tool |
£325.62
Pack of 5 |
In Stock
Log in to see stock. |
|||||
15182769
|
Thermo Scientific™
60180-K106 |
1500 μL | Round | Vial, Mat |
£374.87
Pack of 5 |
In Stock
Log in to see stock. |
|||||
15192769
|
Thermo Scientific™
60180-K107 |
1000 μL | Constricted | Vial, Mat |
£81.86
Each |
In Stock
Log in to see stock. |
|||||
15112779
|
Thermo Scientific™
60180-K109 |
1000 μL | Constricted | Vial, Mat, Plug Cap | N/A | ||||||
Description
Save time and minimize the risk of cross-contamination with Thermo Scientific™ WebSeal™ 96-Well Plate Kits with Sealing Mats. Compatible with all 96-well-configured autosamplers, the ready-to-use kits consist of a comprehensive choice of 96-well microplates, pre-inserted glass or PTFE vials and silicone/PTFE sealing mats.
- Convenient wellplate kits include pre-inserted glass or PTFE vials and 96-position sealing mats; some include cutting tool
- Mats seal samples for storage
- Plates are glass-inserted for greater inertness compared to pure PP plates
- Unique cutting tool allows removal of individual samples with the seal in place, eliminating cross-contamination
Specifications
Standard | |
Webseal | |
96 |
8 x 12 | |
Glass Inserted Plate Kit with 96-Position Mat |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.