missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) A/E Glass Fiber Disc Filters

Superior grade filters for a variety of biological and environmental methods.

Brand:  Cytiva (formerly Pall Lab) 61631

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 17174521

  • £67.65 / Pack of 100
Estimated Shipment: 12-08-2024
to see stock.
Add to basket

Description

Description

  • Type A/E meets the requirements for suspended solids testing, as described in Standard Methods for the Examination of Water and Wastewater, current edition.
  • Reduces filtration costs and premature clogging when filtering difficult-to-filter or highly contaminated solutions.
Specifications

Specifications

60 L/min/cm2
1 μm
100/Pk.
47 mm
Borosilicate glass without binder
Non-sterile
550°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers