missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ AAV-MAX Lysis Buffer

Gibco AAV-MAX Lysis Buffer is a ready-to-use cell lysis reagent for the extraction of adeno-associated virus (AAV) particles from producer HEK293 cells. Cell lysis is detergent-based, eliminating the need for mechanical cell disruption.

Brand:  Gibco™ A50520

125.40 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.


Product Code. 17331899

  • £139.00 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • A ready-to-use cell lysis reagent for the extraction of adeno-associated virus (AAV) particles from producer HEK293 cells
  • Cell lysis is detergent-based, eliminating the need for mechanical cell disruption
  • This provides a milder and easier alternative when isolating AAV particles from cell cultures
  • The buffer is supplied as a 10X solution that can be directly added to HEK293 AAV production cultures to induce cell lysis
  • Gibco AAV-MAX Lysis Buffer is a Polysorbate 20-based formulation and does not contain Triton™ X-100
TRUSTED_SUSTAINABILITY
Specifications

Specifications

10X
100mL Bottle, Store at 2°C to 8°C
Liquid
AAV Production
1 L of Culture
Polysorbate 20 based
Cell Lysis Buffer
Wet Ice
AAV-MAX
AAV
100 mL
6–8
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3048850 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberA50520
Lot Number3010931 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberA50520
Lot Number3025220 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberA50520
Lot Number3086940 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberA50520
Lot Number2891259 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberA50520
Special Offers

Special Offers

  • Multiple Manufacturers
    Advance your proteomics research with essential tools
    Promo Code: 25977

    Expires2025-05-31

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ AAV-MAX Lysis Buffer > 100 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.