missing translation for 'onlineSavingsMsg'
Learn More
Cytiva Ab SpinTrap™ Columns Product Code.: 11703329

Cytiva Ab SpinTrap™ Columns

Product Code. 11703329 Shop All Cytiva Products
50 x 100μL
Click to view available options
:
50 x 100μL
This item is not returnable. View return policy

Product Code. 11703329

Brand: Cytiva 28408347

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use for small-scale purification of IgG. Cytiva Life Sciences™ Ab SpinTrap™ Columns provide a rapid and convenient platform.

No filtration or centrifugation of different crude samples necessary for antibody purification.

  • Protein G Sepharose™ High Performance in prepacked spin columns
  • Binds >1mg human IgG/column
  • No filtration or centrifugation of different crude samples necessary for antibody purification
  • Classic and crosslink protocols for protein enrichment (immunoprecipitation)
  • Elution conditions formatted for both LC-MS and electrophoresis analysis
  • Binding capacity >1mg IgG/column (determined with human polyclonal IgG)
  • Stable in all commonly used buffers within antibody purification
  • Contains polypropylene barrel and polyethylene frits

  • Average particle size: 34μm
  • Bed volume: 100μL
  • Maximum sample volume: 600μL
  • Short term (2h) pH stability: 2–9
  • 600μL undiluted serum sample

TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 100 mL
Binding Capacity > 1 mg IgG/column
Capacity (Metric) 600 μL
Description Ab SpinTrap columns are prepacked, single-use spin columns for rapid purification of monoclonal and polyclonal antibodies from serum and cell culture supernatants. Designed for small-scale purification of multiple samples in parallel.
Support Type 6% Highly Cross-linked Agarose
Content And Storage 4 to 8°C, 20% Ethanol
Quantity 50 x 100 μL
For Use With (Application) Centrifugation
Type Spin Column
Sample Volume 600 μl
compliance-icons compliance-icons
Product Identifier
  • Ab SpinTrap, 50 columns
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P243-Take precautionary measures against static discharge.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva Ab SpinTrap™ Columns >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.