missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AbC™ Total Antibody Compensation Bead Kit
Click to view available options
Quantity:
100 tests
25 tests
Unit Size:
100 tests
25 tests
Description
- Broadest multispecies reactivity—for use with all isotypes of mouse, rat, hamster, and rabbit antibody conjugates
- Brightest signal—most sensitive antibody-capture beads available
- Maximum laser compatibility—for use with all lasers
- Each kit provides polystyrene microspheres that have fluorescent antibody conjugate capture capacity (positive compensation beads) or are inert (negative compensation beads).
- Ideal for compensation with all isotypes of mouse (IgG1, IgG2a, IgG2b, IgG3), rat (IgG1, IgG2a, IgG2b), hamster (Armenian IgG and Syrian IgG), and rabbit (monoclonal and polyclonal) immunoglobulins.Demonstrates the highest and most consistent reactivity to the different subclasses of immunoglobulin in the industry.
Specifications
Specifications
| Diameter (Metric) | 6 μm |
| Content And Storage | Contains 1 vial AbC™ Total Compensation capture beads and 1 vial of negative beads. |
| Format | Tube |
| For Use With (Equipment) | Flow Cytometer |
| No. of Tests | 100 Tests |
| Quantity | 100 tests |
| Detection Method | Fluorescence |
| Excitation Wavelength Range | any lasers (UV to 633/635) |
| Shipping Condition | Room Temperature |
| Product Line | AbC |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction