missing translation for 'onlineSavingsMsg'
Learn More

Multiroir Controlec™ Tilting Trays for ABS Storage Unit

Tilt and/or remove the Multiroir Controlec™ Tilting Trays for ABS Storage Unit for a flexible and versatile arrangement of your items to be stored.

£98.18 - £127.00

Specifications

Color Transparent
Material ABS
Shape Rectangular
For Use With (Equipment) Removable tilt-able drawer unit
Product Type Tilt Bin
View More Specs

Products 2
Product Code Brand Dimensions (L x W x H) Price Quantity & Availability  
Product Code Brand Dimensions (L x W x H) Price Quantity & Availability  
15178042
View Documents
Multiroir Controlec™
50003
197x600x240 mm
£127.00
Each
Please to purchase this item. Need a web account? Register with us today!
11744581
View Documents
Multiroir Controlec™
50003
197x600x240 mm N/A N/A N/A
Description

Description

  • Storage unit is stackable
  • Tilting tray material: ABS
  • Tilting trays in various sizes
Specifications

Specifications

Transparent
Rectangular
Tilt Bin
ABS
Removable tilt-able drawer unit
600 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Multiroir Controlec™ Tilting Trays for ABS Storage Unit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.