missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ AbSelect™ Generic Antibody Purification Kit

Fast and simple method to purify antibodies from ascites fluid and serum

Brand:  Novus Biologicals™ 860-0010

Additional Details : Weight : 0.58200kg

 View more versions of this product

Product Code. 18005765

  • £703.84 / 3 tests
Estimated Shipment: 19-08-2024
to see stock.

Description

Description

The AbSelect method involves capture of the antibody on protein A resin and the removal of unwanted substances by a simple wash procedure. The purified product is then eluted and neutralized. The antibody to be purified or cleaned up ideally is in a volume of 100μL to 0.5mL, up to 500μg of antibody can be purified in each run.

Compatible with lightning-link antibody labeling system, which allows purified antibody to be rapidly conjugated with wide variety of labels

Recommended Storage

Store at 4°C. Do not freeze.

Specifications

Specifications

Antibody Purification Kit
3 Purifications
4°C. Do not freeze.
3 Tests
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers