missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ AbSelect™ Generic Antibody Purification Kit
Fast and simple method to purify antibodies from ascites fluid and serum
£387.33 - £703.84
Specifications
Product Type | Antibody Purification Kit |
---|---|
Content And Storage | 4°C. Do not freeze. |
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
18048685
|
Novus Biologicals™
860-0005 |
1 Purification |
£387.33
1 test |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18005765
|
Novus Biologicals™
860-0010 |
3 Purifications |
£703.84
3 tests |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The AbSelect method involves capture of the antibody on protein A resin and the removal of unwanted substances by a simple wash procedure. The purified product is then eluted and neutralized. The antibody to be purified or cleaned up ideally is in a volume of 100μL to 0.5mL, up to 500μg of antibody can be purified in each run.
Compatible with lightning-link antibody labeling system, which allows purified antibody to be rapidly conjugated with wide variety of labelsSpecifications
Antibody Purification Kit | |
4°C. Do not freeze. |