Learn More
Thermo Scientific™ ABsolute QPCR Mix, SYBR Green, ROX
Optimized for SYBR Green chemistry and contains all the components necessary to perform quantitative PCR, with the exception of template and primers.
£576.00 - £2065.00
Specifications
Detection Method | SYBR |
---|---|
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | Taq DNA Polymerase |
Reaction Speed | Standard |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
10137693
|
Thermo Scientific™
AB1162B |
1,600 Reactions |
£2065.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11872043
|
Thermo Scientific™
AB1163A |
400 Reactions |
£576.00
400 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The 2X qPCR SYBR Green Mix contains optimal levels of active SYBR Green I dye and Thermo-Start DNA Polymerase supplied in a proprietary reaction buffer that enables detection of low copy number targets.
ABsolute qPCR SYBR Green Mixes are available with the option of included high ROX reference dye concentration, Low ROX concentration, Fluorescein or no reference dye. A master mix optimized for capillary instruments is also available.
- High Sensitivity & Specificity - The master mixes contain the patented hot start enzyme Thermo-Start DNA Polymerase
- Optimized buffer containing SYBR Green
- Reproducibility - All master mixes undergo extensive QC to ensure batch to batch consistency
- Broad dynamic range
Specifications
SYBR | |
qPCR | |
Standard | |
ABsolute | |
Gene Expression, Genotyping, siRNA Validation | |
2X |
High | |
Taq DNA Polymerase | |
7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System | |
Real Time PCR SYBR Master Mix | |
qPCR Cyclers | |
DNA, dsDNA |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.