Learn More
Tocris Bioscience™ AC 710
Potent and selective PDGFR family inhibitor
Brand:  Tocris Bioscience™ 5013/10
Additional Details : CAS Number : 1351522-04-7 Weight : 0.00970kg
 View more versions of this product
Product Code. 18720278
  • £241.89 / 10mg
Estimated Shipment: 29-07-2024
to see stock.
Description
Description
AC 710 is a potent PDGFR family inhibitor (IC50 values are 1.2, 2, 7.7 and 10.5 nM for KIT, FLT3, PDGFRβ and CSF1R respectively; Kd = 1.3 nM for PDGFRα). Exhibits >30-fold selectivity over a panel of other kinases and exhibits no activity against cytochrome P450. Causes tumor regression of leukemia cell xenografts in mice. Also reduces joint swelling and inflammation in a mouse model of collagen-induced arthritis. Orally active.
Specifications
Specifications
>99%
562.7
10 mg
AC 710
AC 710