missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AC 710
Potent and selective PDGFR family inhibitor
£241.89 - £950.30
Specifications
Purity | >99% |
---|---|
Description | AC 710 |
Molecular Weight (g/mol) | 562.7 |
Product Type | AC 710 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18720278
|
Tocris Bioscience™
5013/10 |
10 mg |
£241.89
10mg |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18796119
|
Tocris Bioscience™
5013/50 |
50 mg |
£950.30
50mg |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Potent PDGFR family inhibitor (IC50 values are 1.2, 2, 7.7 and 10.5 nM for KIT, FLT3, PDGFRβ and CSF1R respectively; Kd = 1.3 nM for PDGFRα). Exhibits >30-fold selectivity over a panel of other kinases and exhibits no activity against cytochrome P450. Causes tumor regression of leukemia cell xenografts in mice. Also reduces joint swelling and inflammation in a mouse model of collagen-induced arthritis. Orally active.Specifications
>99% | |
AC 710 | |
562.7 | |
AC 710 |
For Research Use Only