Learn More
Brady™ AC Adaptor
AC Adaptor for the BMP™ Label Printers and Label Makers.
Brand:  Brady™ 710613
 View more versions of this product
Product Code. 11751086
  • £190.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 800747: AC Adapter Europe (for ABC or KEY printers).
  • 710612: Accessory for the BMP™41/51/53/61/71 Label Printers.
  • 710613: Accessory for the BMP™51/53 Label Makers.
Specifications
Specifications
AC Adapter
AC Adaptor, UK 240 V
240 V
BMP41 Label Printer, BMP51 Label Printer, BMP53 Label Printer, BMP71 Label Printer, BradyPrinter M611
1 Ea.