missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ AC Adaptor
AC Adaptor for the BMP™ Label Printers and Label Makers.
£178.00 - £191.00
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11820885
|
Brady™
710612 |
AC Adaptor, Europe 220 V |
£191.00
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||||
11751086
|
Brady™
710613 |
AC Adaptor, UK 240 V |
£178.00
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||||
Description
- 800747: AC Adapter Europe (for ABC or KEY printers).
- 710612: Accessory for the BMP™41/51/53/61/71 Label Printers.
- 710613: Accessory for the BMP™51/53 Label Makers.
Specifications
AC Adapter | |
1 Ea. |