missing translation for 'onlineSavingsMsg'
Learn More

Brady™ AC Adaptor

AC Adaptor for the BMP™ Label Printers and Label Makers.

Brand:  Brady™ 710612

181.45 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.



Code : NEW

Additional Details : Weight : 0.59900kg

 View more versions of this product

Product Code. 11820885

  • £191.00 / Each
Estimated Shipment: 06-09-2024
to see stock.
Add to basket

Description

Description

  • 800747: AC Adapter Europe (for ABC or KEY printers).
  • 710612: Accessory for the BMP™41/51/53/61/71 Label Printers.
  • 710613: Accessory for the BMP™51/53 Label Makers.
Specifications

Specifications

AC Adapter
BMP41 Label Printer, BMP51 Label Printer, BMP53 Label Printer, BMP61 Labeling System, BMP71 Label Printer, BradyPrinter M611, BMP41 Label Printer
1 Ea.
Black
AC Adaptor, Europe 220 V
220 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers