Learn More
Thermo Scientific™ Accessories for Cimarec+™ Stirrer Series
Get precise stirring control for your routine protocols.
£17.29 - £55.77
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
15483660
|
Thermo Scientific™
CIC0001448 |
PT1000 External probe 0-400C |
£55.77
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
15413660
|
Thermo Scientific™
CIC0001444 |
Transparent shield for 4x4 |
£20.09
Each |
Estimated Shipment: 04-09-2025
Log in to see stock. |
||||||
15443660
|
Thermo Scientific™
CIC0001445 |
Transparent shield for 7x7 |
£17.29
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||||
15453660
|
Thermo Scientific™
CIC0001446 |
Transparent shield for 10x10 |
£21.36
Each |
Estimated Shipment: 12-03-2026
Log in to see stock. |
||||||
15626245
|
Thermo Scientific™
CIC0001830 |
Supporting Rod |
£26.70
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||||
15636245
|
Thermo Scientific™
CIC0001831 |
Clamp and holder |
£31.96
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||||
Description
These accessories are for use with the Cimarec+™ series digital stirrers. Our new Thermo Scientific™ Cimarec+™ series digital stirrers offer precise stirring control for your routine protocols. The Cimarec+ Series are available in three sizes, providing flexibility from microscale chemistries to production operation.
Advanced technology
- Stir-Trac features innovative engineering for slow-speed stirring, consistent speed control, and strong magnetic coupling
Flexible, robust design
- Top plate choices in ceramic or aluminum
- Easy-to-read LED display for stirring
- Raised display design protects electronics from spills
- Optional splash guard protection shield
- Variable sizes for different sample volumes
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
LED |