Learn More
Thermo Scientific™ Accessories for Cimarec+™ Stirrer Series
Get precise stirring control for your routine protocols.
Brand:  Thermo Scientific™ CIC0001448
45.15 GBP valid until 2025-04-30
Use promo code "24119" to get your promotional price.
 View more versions of this product
Product Code. 15483660
  • £55.77 / Each
Estimated Shipment: 03-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
These accessories are for use with the Cimarec+™ series digital stirrers. Our new Thermo Scientific™ Cimarec+™ series digital stirrers offer precise stirring control for your routine protocols. The Cimarec+ Series are available in three sizes, providing flexibility from microscale chemistries to production operation.
Advanced technology
  • Stir-Trac features innovative engineering for slow-speed stirring, consistent speed control, and strongmagnetic coupling
Flexible, robust design
  • Top plate choices in ceramic or aluminum
  • Easy-to-read LED display for stirring
  • Raised display design protects electronics from spills
  • Optional splash guard protection shield
  • Variable sizes for different sample volumes
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LED
Certificate of Analysis (COA), Certificate of Conformance (COC)
PT1000 External probe 0-400C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Accessories for Cimarec+™ Stirrer Series >