Learn More
Fisherbrand™ Accessories for Fisherbrand™ Heating/Cooling Mini Shaker
Optional accessories to enhance the Fisher Scientific Heating/Cooling Mini Shaker
Brand:  Fisherbrand™
Additional Details : Weight : 0.87000kg
 View more versions of this product
Product Code. 15584080
  • £83.30 / Each
Estimated Shipment: 19-12-2024
to see stock.
Add to basket
Specifications
20 x 1.5 mL micro centrifuge tube block; Well: 11.1mm dia X 39.1mm depth