missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Accessories for Microplate Reader
GREENER_CHOICE

For use with Multi-mode, absorbance and monochromator based microplate readers.

Brand:  Agilent BioTek 7082225

 View more versions of this product

Product Code. 12586746

  • £698.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Serial Cable (BT75034)

  • Modem (9-pin female to 9-pin male)
  • For use with ELx800, Synergy 2, Synergy 2 Alpha, Synergy 2 SL, Synergy H1m, Synergy HT, Synergy Mx, Synergy MxF
Serial Cable (BT75053)
  • Null Modem (9-pin female to 25-pin female)
  • For use with ELx808, FLx800, PowerWave, Precision, Precision XS, Synergy HT
Fluorescence Filter (BT7082225)
  • 590/20 nm
  • For use with Cytation3, FLx800, Synergy 2, Synergy H1, Synergy H4, Synergy HT
Fuse (BT46024)
  • 250V 1.5A
  • For use with ELx808
Fuse (BT46023)
  • 0.75A
  • For use with PowerWave X

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Fluorescence Filter
Agilent BioTek Microplate Readers
590/20 nm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.