missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek Accessories for Microplate Reader
For use with Multi-mode, absorbance and monochromator based microplate readers.
£53.85 - £680.00
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
12586746
|
Agilent BioTek
7082225 |
Fluorescence Filter |
£680.00
Each |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16572392
|
Agilent BioTek
75034 |
Serial Cable; Modem (9-pin female to 9-pin male) |
£56.00
Each |
Estimated Shipment: 24-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11573505
|
Agilent BioTek
75053 |
Serial Cable; Null Modem (9-pin female to 25-pin female) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Serial Cable (BT75034)
- Modem (9-pin female to 9-pin male)
- For use with ELx800, Synergy 2, Synergy 2 Alpha, Synergy 2 SL, Synergy H1m, Synergy HT, Synergy Mx, Synergy MxF
- Null Modem (9-pin female to 25-pin female)
- For use with ELx808, FLx800, PowerWave, Precision, Precision XS, Synergy HT
- 590/20 nm
- For use with Cytation3, FLx800, Synergy 2, Synergy H1, Synergy H4, Synergy HT
- 250V 1.5A
- For use with ELx808
- 0.75A
- For use with PowerWave X
Specifications
Agilent BioTek Microplate Readers |