Learn More
Merck Accessories for Samplicity™ Filtration Systems
To be used with the Samplicity™ G2 Filtration System.
Brand:  Merck SAMTUBING
 View more versions of this product
Product Code. 12910265
  • £110.00 / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
To be used with the Samplicity™ G2 Filtration System.
Powered by vacuum, the system takes minimal manual effort to filter 1 to 8 samples into standard HPLC vials in seconds, with higher recovery than syringe filters provide.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Samplicity G2 Filtration System
Non-sterile
1 Ea.
Tube Set Assembly
Silicone
Analytical sample prep; High Performance Liquid Chromatography (HPLC)
1 Ea.