Learn More
Merck Accessories for Samplicity™ Filtration Systems
To be used with the Samplicity™ G2 Filtration System.
£102.00 - £210.00
Specifications
For Use With (Application) | Analytical sample prep; High Performance Liquid Chromatography (HPLC) |
---|---|
Quantity | 1 Ea. |
Product Code | Brand | Type | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Material | Price | Quantity & Availability | ||||
16641744
|
Merck
SAMP2LID |
Replacement Lid | Thermoplastic Elastomer |
£210.00
Each |
Estimated Shipment: 21-11-2024
Log in to see stock. |
||||
12910265
|
Merck
SAMTUBING |
Tube Set Assembly | Silicone |
£102.00
Each |
Estimated Shipment: 20-11-2024
Log in to see stock. |
||||
Description
To be used with the Samplicity™ G2 Filtration System.
Powered by vacuum, the system takes minimal manual effort to filter 1 to 8 samples into standard HPLC vials in seconds, with higher recovery than syringe filters provide.Specifications
Analytical sample prep; High Performance Liquid Chromatography (HPLC) | |
1 Ea. |