Learn More
Merck Accessories for Samplicity™ Filtration Systems
To be used with the Samplicity™ G2 Filtration System.
Brand: Merck SAMP2LID
This item is not returnable.
View return policy
Description
To be used with the Samplicity™ G2 Filtration System.
Powered by vacuum, the system takes minimal manual effort to filter 1 to 8 samples into standard HPLC vials in seconds, with higher recovery than syringe filters provide.Specifications
Clear | |
Samplicity G2 Filtration System with Millex Filters | |
Analytical sample prep; High Performance Liquid Chromatography (HPLC) | |
1 Ea. | |
Replacement Lid |
Nonsterile lid with doors for attachment of Millex filters/funnels to HPLC tubes | |
Thermoplastic Elastomer | |
PVDF | |
Up to 8 samples |