missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Support Frame (172 mm high with casters)
For Use With: Thermo Scientific Heracell VIOS 160i AND Thermo Scientific Forma Steri-Cycle i160 Incubators
Brand: Thermo Scientific™ 50145394
This item is not returnable.
View return policy
Description
Replacement filter and stand

Specifications
Support frame for double chamber, 172 mm high (with castors) | |
Accessory, Support Frame |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Support Frame (172 mm high with casters) > Support frame for double chamber, 172mm high (with casters)
Spot an opportunity for improvement?Share a Content Correction