Learn More
IKA™ Accessory Stirring Shafts for SteadyStir™ and Overhead Stirrers
Generate axial flow in the vessel with these stirring shafts. IKA™ Accessory Stirring Shafts for SteadyStir™ and IKA™ Overhead Stirrers are designed for medium to high speeds.
Brand:  IKA™ 0002332900
 View more versions of this product
Product Code. 10160161
  • £227.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For drawing material to be mixed from above
  • Generates axial flow in the vessel
  • Minimum danger of injury when contact is made with vessel
  • Minimum shearing forces
  • Use at medium to high speeds
† PTFE-coated
Specifications
Specifications
R 1311
Turbine Stirrer