Learn More
BRAND™ accu-jet™ S Pipet Controller Accessories
Accessories for use with the accu-jet™ S Pipet Controller.
Brand: BRAND™ 26508
Code : 03
Additional Details : Weight : 0.00900kg
Description
BrandTech™ accu-jet™ S Pipet Controller Accessories available for order include:
- Sterile 0.2 μm replacement membrane filters
- Non-Sterile 0.2 μm replacement membrane filters
- Silicone adapter with PTFE check valve
Specifications
accu-jet S | |
Silicone Adapter | |
Each |
Silicone | |
accu-jet™ S Pipet Controller |