Learn More
Fisherbrand™ ACCU500 Water Purification System
Produces ultrapure and reverse osmosis water from tap water directly with no additional pre-treatment filters.
Brand:  Fisherbrand™ RD0P2100EK
Product Code. 15489699
  • £8070.00 / Each
Estimated Shipment: 22-10-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Quality of ultrapure water meets or exceeds ASTM, CLSI, CAP and ISO Type I water standards
  • System CE and RoHS certified
  • Remote dispenser with built-in high precision resistivity sensor with cell constant of 0.01 cm-1 to measure ultrapure water quality just prior to the dispensing point
  • Two years warranty
Specifications
Specifications
ASTM, CLSI, CAP, ISO Type I
<0.001 EU/mL with a terminal ultrafiltration cartridge
1.5 L/min. (ultrapure water type I)
Optional
<1/mL (with a 0.2μm final filter or terminal ultrafiltration cartridge)
30 x 51 x 48 cm
<20µS/cm
Tap, TDS < 1000 ppm (2000μS/cm)
Type 1 and 3
18.2 Mohm.cm
10, 15, 20, 30 L/hr.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ ACCU500 Water Purification System >