Learn More
Hirschmann™ Glass Coverslips for Haemacytometer
Cover glasses for Haemacytometer with smooth surface grinding, without CE mark.
Brand:  Hirschmann™ 8000124
 View more versions of this product
Product Code. 10099780
Description
Description
  • Surface ground and polished surface
  • LOT number on the packaging
  • Labelled with: usable until, CE pursuant to IVD 98/79 EC According to the German Calibration Regulation (Eichordnung), only officially calibrated haemocytometer glass covers may be placed on the market and used in the FRG
Specifications
Specifications
Hemacytometer Coverslip
0.4 mm
Haemocytometer
Glass
20 x 26 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hirschmann™ Glass Coverslips for Haemacytometer > Dimensions: 20 x 26cm