missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AccuCheck Counting Beads

Product Code. 10110533
Click to view available options
Quantity:
10 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10110533

Brand: Invitrogen™ PCB100

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

AccuCheck Counting Beads

AccuCheck Counting Beads are an efficient single-platform method for absolute cell counts that combines the advantages of direct flow cytometric immunophenotyping with the use of two different fluorescent beads (A and B beads). These two fluorospheres are used as a double internal standard for blood volume calculation. A known volume of AccuCheck Counting Beads is added to the same known volume of stained blood in a lyse-no-wash technique. The beads are counted along with cells. Since the concentration of beads is known, the number of cells per microliter (the absolute count) is obtained by relating the number of cells counted to the total number of fluorescent bead events. The cell number is then multiplied by the number of total fluorospheres per unit of volume. As the AccuCheck Counting Beads system contains two different fluorospheres in a known proportion, we can first assure the accuracy of the assay by verifying the proportion of both types of beads.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 6.36 μm, 6.4 μm
Content And Storage Contains 1 bottle of AccuCheck counting beads (10 mL). Store in refrigerator (2–8°C).
Emission 488 and 633/635
Format Solution
For Use With (Equipment) Flow Cytometer
No. of Tests 100 tests
Quantity 10 mL
Detection Method Fluorescence
Excitation Wavelength Range Bead A: 488/575-585 nm, Bead B: 635/660-680 nm
Shipping Condition Wet Ice
Product Type Cell Counting Bead
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.