Learn More
Fisherbrand™ AccuDuo Cartridge
Ensures optimum quality of the final product water. Fisher Scientific™ AccuDuo Cartridges provide dual column filtration for Accu20, Accu100, and Accu500 water systems.
Brand: Fisherbrand™ RR600Q301
Description
- Includes two sets of dual-column purification cartridges
- Cartridges work sequentially to ensure thorough removal of ions and organics in water
Specifications
Accu20, Accu500 | |
U Pack (low TOC) | |
Ultrapure water for critical laboratory applications |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Fisherbrand™ AccuDuo Cartridge >
Spot an opportunity for improvement?Share a Content Correction