Learn More
Fisherbrand™ AccuDuo Cartridge
Ensures optimum quality of the final product water. Fisher Scientific™ AccuDuo Cartridges provide dual column filtration for Accu20, Accu100, and Accu500 water systems.
Brand:  Fisherbrand™ RR600Q201
 View more versions of this product
Product Code. 15489729
  • £330.00 / Each
Estimated Shipment: 30-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Includes two sets of dual-column purification cartridges
  • Cartridges work sequentially to ensure thorough removal of ions and organics in water
Specifications
Specifications
Accu20, Accu500
U Pack
Ultrapure water for critical laboratory applications
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ AccuDuo Cartridge >